6-Bromo-4-fluoro-1H-indazole 97%

Supplier: APOLLO SCIENTIFIC

Danger

PC200355-1G PC200355-250MG PC200355-5G
76617-804EA 40 USD
76617-804 76617-806 76617-802
6-Bromo-4-fluoro-1H-indazole 97%
6-Bromo-4-fluoro-1H-indazole
Formula: C₇H₄BrFN₂
MW: 215.0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781444
CAS Number: 885520-23-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR