6-(Trifluoromethyl)imidazo[1,2-a]pyridine

Supplier: APOLLO SCIENTIFIC
Danger

PC200102-250MG PC200102-5G PC200102-1G
76617-800EA 120 USD
76617-800 76617-796 76617-798
6-(Trifluoromethyl)imidazo[1,2-a]pyridine
6-(Trifluoromethyl)imidazo[1,2-a]pyridine
Formula: C₈H₅F₃N₂
Melting Pt: 98…99 °C
Storage Temperature: Ambient
MDL Number: MFCD09994631
CAS Number: 936009-02-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR