2-Amino-5-chloro-3-fluoropyridine 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 5-Chloro-3-fluoropyridin-2-amine5-Chloro-3-fluoro-2-pyridylamine

PC1964-5G PC1964-25G PC1964-100G
76617-790EA 205 USD
76617-790 76617-792 76617-794
2-Amino-5-chloro-3-fluoropyridine 95%
2-Amino-5-chloro-3-fluoropyridine
Formula: C₅H₄ClFN₂
MW: 146.55 g/mol
Melting Pt: 91…95 °C
Storage Temperature: Ambient
MDL Number: MFCD09878434
CAS Number: 246847-98-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR