5-Chloro-2-nitrobenzotrifluoride 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Chloro-4-nitro-3-(trifluoromethyl)benzene4-Chloro-1-nitro-2-(trifluoromethyl)benzene5-Chloro-2-nitrobenzotrifluoride5-Chloro-α,α,α-trifluoro-2-nitrotoluene

PC1960-100G PC1960-25G
76617-786EA 81.37 USD
76617-786 76617-788
5-Chloro-2-nitrobenzotrifluoride 99%
5-Chloro-2-nitrobenzotrifluoride
Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
Boiling Pt: 222…224 °C
Melting Pt: 21…22 °C
Density: 1.526 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00007298
CAS Number: 118-83-2
UN: 3082
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR