1-Chloro-2-iodotetrafluoroethane 97%

Danger

PC1933L-250MG
76612-950EA 13.39 USD
76612-950
1-Chloro-2-iodotetrafluoroethane 97%
1-Chloro-2-iodotetrafluoroethane
Formula: C₂ClF₄I
Boiling Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00069092
CAS Number: 421-78-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR