α,2-Dichloro-6-fluorotoluene 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 1-Chloro-2-(chloromethyl)-3-fluorobenzene2-Chloro-6-fluorobenzyl chloride

PC1864H-100G PC1864H-250G PC1864H-25G
76622-450EA 15 USD
76622-450 76622-448 76622-446
α,2-Dichloro-6-fluorotoluene 97%
α,2-Dichloro-6-fluorotoluene
Formula: C₇H₅Cl₂F
MW: 179.02 g/mol
Boiling Pt: 208…210 °C
Density: 1.401 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000896
CAS Number: 55117-15-2
EINECS: 259-487-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR