4-Chloro-α,α,α-trifluorotoluene 99%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 4-Chlorobenzotrifluoride1-Chloro-4-(trifluoromethyl)benzene

PC1710-500G
76612-878EA 104 USD
76612-878
4-Chloro-α,α,α-trifluorotoluene 99%
4-Chloro-α,α,α-trifluorotoluene
Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Boiling Pt: 136…139 °C
Melting Pt: –36 °C
Density: 1.353 g/cm³ (25 °C)
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00000627
CAS Number: 98-56-6
EINECS: 202-681-1
UN: 2234
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR