4-Bromo-2,6-difluorobenzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

PC1515-25G PC1515-100G PC1515-5G
76625-736EA 22.42 USD
76625-736 76625-734 76625-732
4-Bromo-2,6-difluorobenzonitrile
4-Bromo-2,6-difluorobenzonitrile
Formula: C₇H₂BrF₂N
MW: 218 g/mol
Melting Pt: 79…82 °C
Storage Temperature: Ambient
MDL Number: MFCD01310981
CAS Number: 123843-67-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR