4-Bromo-3-(trifluoromethyl)benzonitrile 97%

Supplier: APOLLO SCIENTIFIC

Warning

PC1337-5G PC1337-25G
76625-706EA 13 USD
76625-706 76625-704
4-Bromo-3-(trifluoromethyl)benzonitrile 97%
4-Bromo-3-(trifluoromethyl)benzonitrile
Formula: C₈H₃BrF₃N
Storage Temperature: Ambient
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR