2-Bromo-α,α,α-trifluorotoluene 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromobenzotrifluoride1-Bromo-2-(trifluoromethyl)benzene

PC1330-100G PC1330-250G
76612-788EA 46 USD
76612-788 76612-786
2-Bromo-α,α,α-trifluorotoluene 99%
2-Bromo-α,α,α-trifluorotoluene
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 167…168 °C
Density: 1.652 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00000373
CAS Number: 392-83-6
EINECS: 206-879-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR