3,5-Difluoro-2-methoxybenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1136-25G PC1136-5G PC1136-1G
76625-688EA 309 USD
76625-688 76625-686 76625-690
3,5-Difluoro-2-methoxybenzonitrile 98%
3,5-Difluoro-2-methoxybenzonitrile
Formula: C₈H₅F₂NO
MW: 169.13 g/mol
Melting Pt: 33…36 °C
Storage Temperature: Ambient
MDL Number: MFCD06409085
CAS Number: 874804-08-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR