4,5-Difluoro-2-methoxybenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1124-1G PC1124-5G PC1124-10G
76625-684EA 108 USD
76625-684 76625-682 76625-680
4,5-Difluoro-2-methoxybenzonitrile 98%
4,5-Difluoro-2-methoxybenzonitrile
Formula: C₈H₅F₂NO
MW: 169.13 g/mol
Melting Pt: 97…100 °C
Storage Temperature: Ambient
MDL Number: MFCD06660346
CAS Number: 425702-28-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR