2-Fluoro-5-(trifluoromethoxy)benzyl bromide

Supplier: APOLLO SCIENTIFIC

Danger

PC1095-5G PC1095-1G
76622-382EA 242 USD
76622-382 76622-380
2-Fluoro-5-(trifluoromethoxy)benzyl bromide
2-Fluoro-5-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD04115890
CAS Number: 86256-24-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR