TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: TBTU

PC0921-500G PC0921-100G PC0921-25G
76612-678EA 14 USD
76612-678 76612-676 76612-680
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)

Peptide coupling reagent.

Formula: C₁₁H₁₆BF₄N₅O
MW: 321.1 g/mol
Melting Pt: 195…205 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00077413
CAS Number: 125700-67-6
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR