4-(Pentafluorothio)benzonitrile

Supplier: APOLLO SCIENTIFIC

Warning

PC0804-1G PC0804-5G
76625-636EA 604 USD
76625-636 76625-634
4-(Pentafluorothio)benzonitrile
4-(Pentafluorothio)benzonitrile
Formula: C₇H₄F₅NS
Melting Pt: 102…105 °C
Storage Temperature: Ambient
MDL Number: MFCD03788521
CAS Number: 401892-85-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR