2,6-Difluoro-3-methylbenzylamine 98%

Supplier: APOLLO SCIENTIFIC

PC0252-1G PC0252-5G PC0252-10G
76612-614EA 171 USD
76612-614 76612-612 76612-610
2,6-Difluoro-3-methylbenzylamine 98%
2,6-Difluoro-3-methylbenzylamine
Formula: C₈H₉F₂N
Storage Temperature: Ambient
MDL Number: MFCD01631410
CAS Number: 261763-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR