2-Nitro-4-(trifluoromethyl)benzyl bromide 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
1-(Bromomethyl)-2-nitro-4-(trifluoromethyl)benzene
PC01500-1G
76622-128EA
56.65
USD
76622-128
2-Nitro-4-(trifluoromethyl)benzyl bromide 97%
2-Nitro-4-(trifluoromethyl)benzyl bromide
Formula:
C₈H₅BrF₃NO₂ MW: 284.03 g/mol Melting Pt: 36…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095413 CAS Number: 162333-02-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...