3-Chloro-4-fluorophenylacetonitrile

Supplier: APOLLO SCIENTIFIC
Danger

PC0137-5G PC0137-100G PC0137-25G
76625-558EA 22 USD
76625-558 76625-556 76625-554
3-Chloro-4-fluorophenylacetonitrile
3-Chloro-4-fluorophenylacetonitrile
Formula: C₈H₅ClFN
MW: 169.5834 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631553
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR