6-Chloro-2-fluoro-3-methylbenzylamine

Supplier: APOLLO SCIENTIFIC
Danger

PC0113-5G PC0113-10G PC0113-1G
76612-576EA 138 USD
76612-576 76612-574 76612-572
6-Chloro-2-fluoro-3-methylbenzylamine
6-Chloro-2-fluoro-3-methylbenzylamine
Formula: C₈H₉ClFN
Storage Temperature: Ambient
MDL Number: MFCD01631420
CAS Number: 261762-86-1
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR