2-Chloro-6-fluoro-3-methylbenzaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-6-fluoro-3-tolylaldehyde2-Chloro-6-fluoro-m-tolylaldehyde2-Chloro-6-fluoro-3-tolualdehyde2-Chloro-6-fluoro-m-tolualdehyde

PC0093-1G PC0093-5G PC0093-25G
76591-118EA 177 USD
76591-118 76591-114 76591-116
2-Chloro-6-fluoro-3-methylbenzaldehyde 95%
2-Chloro-6-fluoro-3-methylbenzaldehyde
Formula: C₈H₆ClFO
MW: 172.59 g/mol
Melting Pt: 26…33 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01631360
CAS Number: 104451-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR