4-(2-Aminoethyl)benzenesulfonamide

Supplier: APOLLO SCIENTIFIC

Danger

OR9989-25G OR9989-500G OR9989-100G
76617-548EA 205 USD
76617-548 76617-544 76617-546
4-(2-Aminoethyl)benzenesulfonamide
4-(2-Aminoethyl)benzenesulphonamide
Formula: C₈H₁₂N₂O₂S
MW: 200.26 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00010301
CAS Number: 35303-76-5
EINECS: 252-501-0
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR