5-Methoxy-1-indanone 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR9975-5G OR9975-25G OR9975-100G
76590-968EA 81 USD
76590-968 76590-966 76590-970
5-Methoxy-1-indanone 98%
5-Methoxy-1-indanone
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Melting Pt: 107…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003789
CAS Number: 5111-70-6
EINECS: 225-838-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR