2-Methoxy-6-methylpyridine 98%

Supplier: APOLLO SCIENTIFIC

Warning

OR9940-100G OR9940-25G
76612-548EA 173 USD
76612-548 76612-550
2-Methoxy-6-methylpyridine 98%
2-Methoxy-6-methylpyridine
Formula: C₇H₉NO
MW: 123.16 g/mol
Boiling Pt: 156 °C
Density: 1.01 g/cm³ (25 °C)
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD04112507
CAS Number: 63071-03-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR