Ethyl-2,4-dimethylpyrrole-3-carboxylate 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Ethyl 2,4-dimethyl-1H-pyrrole-3-carboxylate

OR9926-1G OR9926-5G
76590-916EA 16 USD
76590-916 76590-918
Ethyl-2,4-dimethylpyrrole-3-carboxylate 98%
Ethyl 2,4-dimethylpyrrole-3-carboxylate
Formula: C₉H₁₃NO₂
MW: 167.2050 g/mol
Melting Pt: 122…123 °C
Storage Temperature: Ambient
MDL Number: MFCD01365811
CAS Number: 2199-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR