3-Amino-4-chloro-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Chloro-1H-indazol-3-amine

OR9905-5G OR9905-25G
76590-898EA 252 USD
76590-898 76590-896
3-Amino-4-chloro-1H-indazole
3-Amino-4-chloro-1H-indazole
Formula: C₇H₆ClN₃
MW: 167.60 g/mol
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00663139
CAS Number: 20925-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR