Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate

OR9878-1G
76590-874EA 32 USD
76590-874
Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate
Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate
Formula: C₈H₁₂N₂O₂
MW: 168.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159638
CAS Number: 35691-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR