2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one 95%

Supplier: APOLLO SCIENTIFIC

Warning

OR9861-5G OR9861-1G OR9861-10G
76590-836EA 52 USD
76590-836 76590-834 76590-838
2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one 95%
2H-Pyrido[3,2-b][1,4]oxazin-3(4H)-one
Formula: C₇H₆N₂O₂
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00006697
CAS Number: 20348-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR