2-Amino-1-(4-chlorophenyl)-1-ethanol 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
alpha-(Aminomethyl)-4-chlorobenzyl alcohol
OR9825-500MG
OR9825-5G
OR9825-1G
76590-818EA
273
USD
76590-818
76590-816
76590-820
2-Amino-1-(4-chlorophenyl)-1-ethanol 95%
2-Amino-1-(4-chlorophenyl)-1-ethanol
Formula:
C₈H₁₀ClNO MW: 171.63 g/mol Melting Pt: 93…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797373 CAS Number: 41870-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...