Ethyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR9789-250MG OR9789-50MG
76590-794EA 18 USD
76590-794 76590-796
Ethyl-2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Ethyl 2-phenylimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₁₆H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00139520
CAS Number: 119448-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR