4-Bromobenzyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: α-Chloro-4-bromotoluene1-(Chloromethyl)-4-bromobenzene4-Bromo-α-chlorotoluene1-Bromo-4-(chloromethyl)benzene

OR9776-25G OR9776-5G
76622-042EA 19.57 USD
76622-042 76622-040
4-Bromobenzyl chloride
4-Bromobenzyl chloride
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 106…107 °C/10mmHg
Melting Pt: 37…41 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00040867
CAS Number: 589-17-3
EINECS: 209-638-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR