3-Pyrid-4-ylbenzoic acid 95%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 3-(Pyridin-4-yl)benzoic acid

OR9726-250MG OR9726-1G
76590-736EA 81 USD
76590-736 76590-734
3-Pyrid-4-ylbenzoic acid 95%
3-Pyrid-4-ylbenzoic acid
Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
Melting Pt: 268 °C
Storage Temperature: Ambient
MDL Number: MFCD05864808
CAS Number: 4385-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR