2-Mercapto-5-methoxybenzimidazole 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Methoxy-1H-benzo[d]imidazole-2-thiol5-Methoxybenzimidazole-2-thiol2-Mercapto-5-methoxy-1H-benzimidazole5-Methoxy-2-benzimidazolethiol1,3-Dihydro-5-methoxy-2H-benzimidazole-2-thione

OR9581-100G OR9581-25G OR9581-500G
76590-536EA 80 USD
76590-536 76590-534 76590-532
2-Mercapto-5-methoxybenzimidazole 98%
2-Mercapto-5-methoxybenzimidazole
Formula: C₈H₈N₂OS
MW: 180.23 g/mol
Melting Pt: 255…263 °C
Storage Temperature: Ambient
MDL Number: MFCD00134581
CAS Number: 37052-78-1
EINECS: 253-326-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR