Methyl-6-bromonicotinate 97%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Methyl 6-bromonicotinate

OR9561-25G OR9561-5G
76590-490EA 68 USD
76590-490 76590-488
Methyl-6-bromonicotinate 97%
Methyl 6-bromonicotinate
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD04972371
CAS Number: 26218-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR