4-Bromo-6-chloro-1H-indazole 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR951968-10G OR951968-5G OR951968-1G
76590-390EA 196 USD
76590-390 76590-388 76590-386
4-Bromo-6-chloro-1H-indazole 98%
4-Bromo-6-chloro-1H-indazole
Formula: C₇H₄BrClN₂
MW: 231.4800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07781352
CAS Number: 885519-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR