2-Chloro-5-(hydroxymethyl)phenylboronic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Chloro-5-hydroxymethylphenylboronic acid

OR9493-25G OR9493-5G OR9493-1G
76590-280EA 110 USD
76590-280 76590-282 76590-278
2-Chloro-5-(hydroxymethyl)phenylboronic acid
2-Chloro-5-(hydroxymethyl)phenylboronic acid
Formula: C₇H₈BClO₃
MW: 186.4000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06656270
CAS Number: 1003042-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR