4-tert-Butyl-1-bromobenzene, Technical Grade
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1-Bromo-4-(tert-butyl)benzene
, 2-(4-Bromophenyl)-2-methylpropane
, 4-Bromo-tert-butylbenzene
, 1-Bromo-4-tert-butylbenzene
OR9457-100G
OR9457-500G
OR9457-50G
76590-222EA
20
USD
76590-222
76590-226
76590-224
4-tert-Butyl-1-bromobenzene, Technical Grade
4-tert-Butyl-1-bromobenzene
Formula:
C₁₀H₁₃Br MW: 213.12 g/mol Boiling Pt: 228…232 °C Melting Pt: 13…16 °C Density: 1.229 g/cm³ (25 °C) Flash Pt: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000108 CAS Number: 3972-65-4 EINECS: 223-599-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...