2-(Hydroxymethyl)phenylboronic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-(Hydroxymethyl)benzeneboronic acid(2-(Hydroxymethyl)phenyl)boronic acid

OR9454-25G OR9454-1G OR9454-5G
76590-218EA 26 USD
76590-218 76590-220 76590-216
2-(Hydroxymethyl)phenylboronic acid
2-(Hydroxymethyl)phenylboronic acid
Formula: C₇H₉BO₃
MW: 151.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02258966
CAS Number: 87199-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR