3-Carboxy-4-chlorophenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Borono-2-chlorobenzoic acid

OR9342-1G
76590-034EA 19.57 USD
76590-034
3-Carboxy-4-chlorophenylboronic acid
3-Carboxy-4-chlorophenylboronic acid
Formula: C₇H₆BClO₄
MW: 200.3800 g/mol
Melting Pt: 223…225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06801689
CAS Number: 913835-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR