3-Amino-2-bromo-5-methoxypyridine 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: 2-Bromo-5-methoxypyridin-3-amine

OR929970-250MG OR929970-1G
76589-936EA 53 USD
76589-936 76589-938
3-Amino-2-bromo-5-methoxypyridine 98%
3-Amino-2-bromo-5-methoxypyridine
Formula: C₆H₇BrN₂O
MW: 203.0366 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16659663
CAS Number: 1043688-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR