4'-Chloro-3'-methylacetophenone 99%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: 1-(4-Chloro-3-methylphenyl)ethanone

OR925182-25G OR925182-5G OR925182-1G
76589-814EA 83 USD
76589-814 76589-810 76589-812
4'-Chloro-3'-methylacetophenone 99%
4'-Chloro-3'-methylacetophenone
Formula: C₉H₉ClO
MW: 168.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00045172
CAS Number: 37074-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR