(R)-(+)-N-Benzyl-α-methylbenzylamine 98%

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: (R)-(+)-N-Benzyl-1-phenylethylamine(R)-N-Benzyl-1-phenylethanamine

OR911995-100G OR911995-25G
76589-586EA 66 USD
76589-586 76589-584
(R)-(+)-N-Benzyl-α-methylbenzylamine 98%
(R)-(+)-N-Benzyl-α-methylbenzylamine
Formula: C₁₅H₁₇N
MW: 211.31 g/mol
Boiling Pt: 171 °C/15mmHg
Density: 1.01 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00015010
CAS Number: 38235-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR