1-Ethyl-4-iodo-1H-pyrazole

Supplier: APOLLO SCIENTIFIC
Warning

OR910150-1G OR910150-5G
76589-538EA 18.54 USD
76589-538 76589-540
1-Ethyl-4-iodo-1H-pyrazole
1-Ethyl-4-iodo-1H-pyrazole
Formula: C₅H₇IN₂
Storage Temperature: Ambient
MDL Number: MFCD13195706
CAS Number: 172282-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR