2,3-Dibromo-5-picoline

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,3-Dibromo-5-methylpyridine

OR8978-5G OR8978-1G
76617-444EA 13 USD
76617-444 76617-442
2,3-Dibromo-5-picoline
2,3-Dibromo-5-picoline
Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Boiling Pt: 270.8 °C
Melting Pt: 52…56 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04112574
CAS Number: 29232-39-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR