8-Quinolinecarboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Quinoline-8-carboxylic acid

OR8958-1G OR8958-5G
76589-256EA 75 USD
76589-256 76589-254
8-Quinolinecarboxylic acid
8-Quinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 183…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00047619
CAS Number: 86-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR