5-Isoquinolinecarboxylic acid

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: Isoquinoline-5-carboxylic acid

OR8925-250MG OR8925-1G
76589-218EA 25 USD
76589-218 76589-220
5-Isoquinolinecarboxylic acid
5-Isoquinolinecarboxylic acid
Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 60…62 °C/11mmHg
Melting Pt: 275…281 °C
Storage Temperature: Ambient
MDL Number: MFCD03788744
CAS Number: 27810-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR