Methyl malonyl chloride 97%

Supplier: APOLLO SCIENTIFIC

Danger

Synonyms: Methyl 3-chloro-3-oxopropanoate

OR8845-100G OR8845-250G OR8822-25G OR8845-25G
76589-118EA 53 USD
76589-118 76621-936 76621-932 76621-934
Methyl malonyl chloride 97%
Methyl malonyl chloride
Formula: C₄H₅ClO₃
MW: 136.53 g/mol
Boiling Pt: 57…59 °C/12mmHg
Density: 1.273 g/cm³ (25 °C)
Flash Pt: 80 °C
Storage Temperature: Refrigerator
EINECS: 253-540-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR