(S)-(-)-4-Benzyl-2-oxazolidinone

Supplier: APOLLO SCIENTIFIC

Warning

Synonyms: (S)-4-Benzyloxazolidin-2-one(S)-4-Benzyl-2-oxazolidinone

OR8747-500G OR8747-25G OR8747-100G
76589-046EA 47 USD
76589-046 76589-048 76589-044
(S)-(-)-4-Benzyl-2-oxazolidinone
(S)-(-)-4-Benzyl-2-oxazolidinone
Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00064496
CAS Number: 90719-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR