2-Bromo-4-tert-butylphenol

Supplier: APOLLO SCIENTIFIC
Warning

OR8716-5G OR8716-100G OR8716-25G
76588-998EA 14.42 USD
76588-998 76589-002 76589-000
2-Bromo-4-tert-butylphenol
2-Bromo-4-tert-butylphenol
Formula: C₁₀H₁₃BrO
MW: 229.1100 g/mol
Boiling Pt: 113 °C/9mmHg
Melting Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD02682891
CAS Number: 2198-66-5
EINECS: 218-602-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR