6-Bromo-1,3-dichloroisoquinoline 96%

Supplier: APOLLO SCIENTIFIC

Danger

OR8568-1G OR8568-5G
76617-374EA 103 USD
76617-374 76617-376
6-Bromo-1,3-dichloroisoquinoline 96%
6-Bromo-1,3-dichloroisoquinoline
Formula: C₉H₄BrCl₂N
MW: 276.9448 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD09258639
CAS Number: 552331-05-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR