α-Bromo-4-chlorotoulene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: alpha-Bromo-4-chlorotoluene4-Chlorobenzyl bromide1-(Bromomethyl)-4-chlorobenzene

OR8476-5G OR8476-25G OR8476-100G
76621-918EA 13.39 USD
76621-918 76621-914 76621-916
α-Bromo-4-chlorotoulene 98%
α-Bromo-4-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 107…108 °C/8mmHg
Melting Pt: 46…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00040714
CAS Number: 622-95-7
EINECS: 210-760-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR